Assay Details
Target Gene Details
Entrez Gene ID: | 83723 |
Gene Name: | family with sequence similarity 57 member B |
Gene Aliases: |
FP1188 |
Location: |
Chr.16:30024423-30053026 on Build GRCh38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM57B | NM_001318504.1 | 5 | 517 | NP_001305433.1 |
| NM_031478.5 | 5 | 955 | NP_113666.2 | |
| XM_005255613.3 | 9 | 1163 | XP_005255670.1 | |
| XM_005255614.3 | 8 | 1103 | XP_005255671.1 | |
| XM_005255615.2 | 8 | 1300 | XP_005255672.1 | |
| XM_017023749.1 | 6 | 1147 | XP_016879238.1 | |
| XM_017023750.1 | 9 | 1360 | XP_016879239.1 | |
| XM_017023751.1 | 10 | 1256 | XP_016879240.1 | |
| XM_017023752.1 | 9 | 1159 | XP_016879241.1 | |
| XM_017023753.1 | 7 | 1239 | XP_016879242.1 | |
| XM_017023754.1 | 6 | 661 | XP_016879243.1 | |
| AB593089.1 | 5 | 895 | ||
| AB593090.1 | 5 | 893 | ||
| AF370365.1 | 5 | 907 | AAQ15201.1 | |
| AL136777.1 | 5 | 517 | CAB66711.1 | |
| BC007892.2 | 5 | 673 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1160384 | Chr.16:30023148 - 30178660 on Build GRCh38 | Duplication |
|
More Information
Set Membership: |
|

Genomic Map