Assay Details
Target Gene Details
Entrez Gene ID: | 117143 |
Gene Name: | transcriptional adaptor 1 |
Gene Aliases: |
ADA1, HFI1, STAF42, TADA1L, hADA1 |
Location: |
Chr.1:166856510-166876417 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TADA1 | NM_053053.3 | 8 | 1874 | NP_444281.1 |
| BC015401.2 | 8 | 1699 | AAH15401.1 |
More Information
Set Membership: |
|

Genomic Map