Assay Details
Target Gene Details
Entrez Gene ID: | 79963 |
Gene Name: | ATP binding cassette subfamily A member 11, pseudogene |
Gene Aliases: |
ABCA11, EST1133530 |
Location: |
Chr.4:425435-474209 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ABCA11P | NR_002451.2 | |||
| AK024359.1 | BAB14895.1 | |||
| AK303674.1 | ||||
| DA501191.1 |
Target Gene Details
Entrez Gene ID: | 170960 |
Gene Name: | zinc finger protein 721 |
Gene Aliases: |
- |
Location: |
Chr.4:439984-499171 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZNF721 | NM_133474.3 | 3 | 4472 | NP_597731.2 |
| AB075862.1 | 1 | 3292 | BAB85568.1 | |
| AI223001.1 | 1 | 225 | ||
| AK095689.1 | 1 | 2369 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv461144 | Chr.4:337951 - 480163 on Build GRCh38 | Loss |
|
| dgv8875n54 | Chr.4:354378 - 440290 on Build GRCh38 | Loss |
|
| nsv1009336 | Chr.4:273074 - 481679 on Build GRCh38 | Gain |
|
| nsv1001899 | Chr.4:68917 - 523947 on Build GRCh38 | Loss |
|
| nsv461145 | Chr.4:421522 - 503349 on Build GRCh38 | Gain |
|
| dgv5077n100 | Chr.4:355495 - 453210 on Build GRCh38 | Loss |
|
| esv3893859 | Chr.4:416226 - 503349 on Build GRCh38 | Gain |
|
| nsv1006781 | Chr.4:343385 - 560798 on Build GRCh38 | Gain |
|
| dgv5078n100 | Chr.4:435303 - 488008 on Build GRCh38 | Loss |
|
| nsv593203 | Chr.4:432092 - 482918 on Build GRCh38 | Loss |
|
| nsv1012061 | Chr.4:412575 - 445782 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map