Assay Details
Target Gene Details
Entrez Gene ID: | 7223 |
Gene Name: | transient receptor potential cation channel subfamily C member 4 |
Gene Aliases: |
HTRP-4, HTRP4, TRP4 |
Location: |
Chr.13:37633872-37870374 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TRPC4 | NM_001135955.1 | NP_001129427.1 | ||
| NM_001135956.1 | NP_001129428.1 | |||
| NM_001135957.1 | NP_001129429.1 | |||
| NM_001135958.1 | 10 | 2037 | NP_001129430.1 | |
| NM_003306.1 | 11 | 2571 | NP_003297.1 | |
| NM_016179.2 | 11 | 2556 | NP_057263.1 | |
| XM_011535206.1 | 8 | 1191 | XP_011533508.1 | |
| XM_017020723.1 | 8 | 1191 | XP_016876212.1 | |
| XM_017020724.1 | 11 | 3117 | XP_016876213.1 | |
| AF063822.1 | 11 | 2556 | AAF22927.1 | |
| AF063823.1 | AAF22928.1 | |||
| AF063824.1 | AAF22929.1 | |||
| AF063825.1 | AAF22930.1 | |||
| AF175406.1 | 11 | 2552 | AAD51736.1 | |
| AF421358.1 | 10 | 2321 | AAL24549.1 | |
| AF421359.1 | AAL24550.1 | |||
| AF421360.1 | 10 | 2336 | AAL24551.1 | |
| AF421361.1 | 9 | 1984 | AAL24552.1 | |
| AF421362.1 | 9 | 1802 | AAL24553.1 | |
| AK312524.1 | 11 | 2526 | ||
| BC104725.1 | 11 | 2477 | AAI04726.1 | |
| BC105306.1 | AAI05307.1 | |||
| BC105641.1 | 11 | 2477 | AAI05642.1 | |
| BC105917.1 | 11 | 2477 | AAI05918.1 | |
| BX648312.1 | 11 | 3180 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv34109 | Chr.13:37269682 - 37702254 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map