Assay Details
Target Gene Details
Entrez Gene ID: | 444 |
Gene Name: | aspartate beta-hydroxylase |
Gene Aliases: |
AAH, BAH, CASQ2BP1, FDLAB, HAAH, JCTN, junctin |
Location: |
Chr.8:61500556-61714640 on Build GRCh38 |
Assay Gene Location: | Within Exon 32 |
Target Gene Details
Entrez Gene ID: | 157807 |
Gene Name: | clavesin 1 |
Gene Aliases: |
CRALBPL, RLBP1L1 |
Location: |
Chr.8:61057158-61501645 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CLVS1 | NM_173519.2 | 6 | 3071 | NP_775790.1 |
| XM_017013141.1 | 7 | 3062 | XP_016868630.1 | |
| XM_017013142.1 | 7 | 3219 | XP_016868631.1 | |
| AK094567.1 | 2 | 3004 | ||
| AK125153.1 | 5 | 2318 | ||
| AY094971.1 | 6 | 3029 | AAM15733.1 | |
| BC042617.1 | 6 | 3046 | AAH42617.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3617390 | Chr.8:61457269 - 61656463 on Build GRCh38 | Gain |
|
| esv3617389 | Chr.8:61453750 - 61594908 on Build GRCh38 | Gain |
|
| nsv521898 | Chr.8:61452104 - 61506561 on Build GRCh38 | Gain |
|
| nsv1029046 | Chr.8:61311782 - 61622718 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map