Assay Details
Target Gene Details
Entrez Gene ID: | 57569 |
Gene Name: | Rho GTPase activating protein 20 |
Gene Aliases: |
RARHOGAP |
Location: |
Chr.11:110577035-110713189 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 19 - Exon 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ARHGAP20 | NM_001258415.1 | NP_001245344.1 | ||
| NM_001258416.1 | NP_001245345.1 | |||
| NM_001258417.1 | NP_001245346.1 | |||
| NM_001258418.1 | NP_001245347.1 | |||
| NM_020809.3 | NP_065860.2 | |||
| XM_005271628.3 | XP_005271685.1 | |||
| XM_006718893.3 | XP_006718956.1 | |||
| AB037812.1 | ||||
| AK292290.1 | ||||
| AK307602.1 | ||||
| AY496263.1 | AAS45466.1 | |||
| AY496264.1 | AAS45467.1 | |||
| AY496265.1 | AAS45468.1 | |||
| AY496266.1 | AAS45469.1 | |||
| AY496267.1 | AAS45470.1 | |||
| BC039340.1 | AAH39340.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs116498873] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map