Assay Details
Target Gene Details
Entrez Gene ID: | 4190 |
Gene Name: | malate dehydrogenase 1 |
Gene Aliases: |
HEL-S-32, MDH-s, MDHA, MGC:1375, MOR2 |
Location: |
Chr.2:63588609-63607197 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MDH1 | NM_001199111.1 | NP_001186040.1 | ||
| NM_001199112.1 | NP_001186041.1 | |||
| NM_001316374.1 | NP_001303303.1 | |||
| NM_005917.3 | NP_005908.1 | |||
| AK295931.1 | ||||
| AK300719.1 | ||||
| AK307273.1 | ||||
| AK312331.1 | ||||
| BC001484.2 | AAH01484.1 | |||
| BI667299.1 | ||||
| CB266998.1 | ||||
| CR457405.1 | CAG33686.1 | |||
| D55654.1 | BAA09513.1 | |||
| EU794635.1 | ||||
| U20352.1 | AAC16436.1 |
Target Gene Details
Entrez Gene ID: | 51057 |
Gene Name: | WD repeat containing planar cell polarity effector |
Gene Aliases: |
BBS15, C2orf86, CHDTHP, FRITZ, FRTZ |
Location: |
Chr.2:63120755-63840825 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| WDPCP | XM_017004251.1 | XP_016859740.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv582126 | Chr.2:63346276 - 64470570 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map