Assay Details
Target Gene Details
Entrez Gene ID: | 93643 |
Gene Name: | tight junction associated protein 1 |
Gene Aliases: |
PILT, TJP4 |
Location: |
Chr.6:43477523-43506557 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv603000 | Chr.6:43397622 - 43524840 on Build GRCh38 | Loss |
|
| esv3346999 | Chr.6:43498736 - 43500434 on Build GRCh38 | Duplication |
|
| nsv950173 | Chr.6:43487563 - 43513862 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map