Assay Details
Target Gene Details
Entrez Gene ID: | 143872 |
Gene Name: | Rho GTPase activating protein 42 |
Gene Aliases: |
GRAF3 |
Location: |
Chr.11:100687676-100993941 on Build GRCh38 |
Assay Gene Location: | Within Exon 29 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ARHGAP42 | XM_011542615.2 | 24 | 9125 | XP_011540917.1 |
| XM_011542616.2 | 24 | 6894 | XP_011540918.1 | |
| XM_011542617.2 | 25 | 6780 | XP_011540919.1 | |
| XM_017017237.1 | 24 | 7083 | XP_016872726.1 | |
| XM_017017238.1 | 25 | 9266 | XP_016872727.1 |
Target Gene Details
Entrez Gene ID: | 83935 |
Gene Name: | transmembrane protein 133 |
Gene Aliases: |
AD031 |
Location: |
Chr.11:100992080-100993935 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TMEM133 | NM_032021.2 | 1 | 731 | NP_114410.1 |
| AF247167.1 | 1 | 731 | AAG44547.1 | |
| BC093843.1 | AAH93843.1 | |||
| BC112050.1 | AAI12051.1 |
More Information
Set Membership: |
|

Genomic Map