Assay Details
Target Gene Details
Entrez Gene ID: | 1595 |
Gene Name: | cytochrome P450 family 51 subfamily A member 1 |
Gene Aliases: |
CP51, CYP51, CYPL1, LDM, P450-14DM, P450L1 |
Location: |
Chr.7:92112149-92134745 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CYP51A1 | NM_000786.3 | NP_000777.1 | ||
| NM_001146152.1 | NP_001139624.1 | |||
| AK295932.1 | ||||
| AK314205.1 | ||||
| BC032322.1 | AAH32322.1 | |||
| D55653.1 | BAA09512.1 | |||
| U23942.1 | AAB39951.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3571987 | Chr.7:92120895 - 92126807 on Build GRCh38 | Loss |
|
| nsv1109898 | Chr.7:92122238 - 92124872 on Build GRCh38 | Deletion |
|
| nsv5839 | Chr.7:92123796 - 92150740 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map