Assay Details
Target Gene Details
Entrez Gene ID: | 85444 |
Gene Name: | leucine rich repeat and coiled-coil centrosomal protein 1 |
Gene Aliases: |
CLERC, SAP2, VFL1 |
Location: |
Chr.8:85107088-85146080 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LRRCC1 | NM_033402.4 | 8 | 1384 | NP_208325.3 |
| XM_005251318.3 | 7 | 1173 | XP_005251375.1 | |
| XM_005251319.4 | 6 | 867 | XP_005251376.1 | |
| XM_005251320.3 | 5 | 659 | XP_005251377.1 | |
| XM_005251322.3 | 4 | 473 | XP_005251379.1 | |
| XM_017013920.1 | 9 | 1332 | XP_016869409.1 | |
| XM_017013921.1 | 9 | 1332 | XP_016869410.1 | |
| XM_017013922.1 | 9 | 1332 | XP_016869411.1 | |
| XM_017013923.1 | 8 | 1159 | XP_016869412.1 | |
| XM_017013924.1 | 9 | 1332 | XP_016869413.1 | |
| XM_017013925.1 | 6 | 878 | XP_016869414.1 | |
| XM_017013926.1 | 6 | 861 | XP_016869415.1 | |
| XM_017013927.1 | 7 | 889 | XP_016869416.1 | |
| XM_017013928.1 | 4 | 341 | XP_016869417.1 | |
| AB051551.1 | 7 | 2005 | BAB21855.1 | |
| AK056185.1 | 7 | 1347 | BAB71115.1 | |
| AK300234.1 | ||||
| AK300874.1 | 7 | 1028 | ||
| AK302648.1 | 8 | 1234 | ||
| AK307350.1 | 4 | 448 | ||
| AK307443.1 | 5 | 640 | ||
| AK307636.1 | 7 | 1018 | ||
| BC030701.2 | 8 | 1257 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv483121 | Chr.8:85023159 - 85193909 on Build GRCh38 | Loss |
|
| nsv483126 | Chr.8:83687766 - 85887771 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map