Assay Details
Target Gene Details
Entrez Gene ID: | 2635 |
Gene Name: | guanylate binding protein 3 |
Gene Aliases: |
- |
Location: |
Chr.1:89006677-89022880 on Build GRCh38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GBP3 | NM_001319179.1 | 12 | 2341 | NP_001306108.1 |
| NM_001319180.1 | 11 | 2397 | NP_001306109.1 | |
| NM_018284.2 | 11 | 2255 | NP_060754.2 | |
| NR_135007.1 | 9 | 1761 | ||
| XM_005270743.2 | 12 | 2331 | XP_005270800.1 | |
| XM_006710574.2 | 11 | 2188 | XP_006710637.1 | |
| XM_011541233.1 | 10 | 2043 | XP_011539535.1 | |
| XM_011541234.1 | 10 | 2026 | XP_011539536.1 | |
| XM_017001013.1 | 10 | 2175 | XP_016856502.1 | |
| XM_017001014.1 | 9 | 1923 | XP_016856503.1 | |
| AK309096.1 | 10 | 2895 | ||
| AL136680.1 | 11 | 2154 | CAB66615.1 | |
| BC140837.1 | 11 | 2140 | ||
| CR936755.1 | 11 | 2256 | ||
| CR936758.1 | 11 | 2196 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2758948 | Chr.1:88946385 - 89095040 on Build GRCh38 | Gain |
|
| nsv546785 | Chr.1:88994245 - 89010744 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map