Assay Details
Target Gene Details
Entrez Gene ID: | 5033 |
Gene Name: | prolyl 4-hydroxylase subunit alpha 1 |
Gene Aliases: |
P4HA |
Location: |
Chr.10:73007217-73096974 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 15 - Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| P4HA1 | NM_000917.3 | NP_000908.2 | ||
| NM_001017962.2 | NP_001017962.1 | |||
| NM_001142595.1 | NP_001136067.1 | |||
| NM_001142596.1 | NP_001136068.1 | |||
| AK222960.1 | BAD96680.1 | |||
| AK312593.1 | ||||
| BC034998.1 | AAH34998.1 | |||
| CD013929.1 | ||||
| CD013930.1 | ||||
| M24486.1 | AAA36534.1 | |||
| M24487.1 | AAA36535.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv528473 | Chr.10:72911853 - 73049838 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map