Assay Details
Target Gene Details
Entrez Gene ID: | 140576 |
Gene Name: | S100 calcium binding protein A16 |
Gene Aliases: |
AAG13, DT1P1A7, S100F |
Location: |
Chr.1:153606883-153613168 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| S100A16 | NM_001317007.1 | NP_001303936.1 | ||
| NM_001317008.1 | NP_001303937.1 | |||
| NM_080388.2 | NP_525127.1 | |||
| NR_133645.1 | ||||
| AK290804.1 | ||||
| AW954845.1 | ||||
| AY221961.1 | AAP46152.1 | |||
| AY762098.1 | ||||
| BC010541.1 | AAH10541.1 | |||
| BC019099.1 | AAH19099.1 | |||
| BC095462.1 | AAH95462.1 | |||
| BM739127.1 | ||||
| BQ439720.1 | ||||
| HY178745.1 | ||||
| U92985.1 |
More Information
Set Membership: |
|

Genomic Map