Assay Details
Target Gene Details
Entrez Gene ID: | 84639 |
Gene Name: | interleukin 1 family member 10 (theta) |
Gene Aliases: |
FIL1-theta, FKSG75, IL-1HY2, IL-38, IL1-theta, IL1HY2 |
Location: |
Chr.2:113067970-113075850 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IL1F10 | NM_032556.5 | 4 | 850 | NP_115945.4 |
| NM_173161.2 | 5 | 504 | NP_775184.1 | |
| AF334755.1 | 4 | 850 | AAK68048.1 | |
| AY026753.1 | 2 | 495 | AAK01948.1 | |
| AY029413.1 | 5 | 504 | AAK33010.1 | |
| AY358846.1 | 5 | 492 | AAQ89205.1 | |
| BC103966.1 | 5 | 495 | AAI03967.1 | |
| BC103967.1 | 5 | 495 | AAI03968.1 | |
| BC103968.1 | 5 | 495 | AAI03969.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv582710 | Chr.2:113074066 - 113075404 on Build GRCh38 | Gain |
|
| nsv2881 | Chr.2:113043228 - 113089020 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map