Assay Details
Target Gene Details
Entrez Gene ID: | 5794 |
Gene Name: | protein tyrosine phosphatase, receptor type H |
Gene Aliases: |
R-PTP-H, SAP1 |
Location: |
Chr.19:55181247-55209506 on Build GRCh38 |
Assay Gene Location: | Within Exon 21 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PTPRH | NM_001161440.2 | 18 | 3185 | NP_001154912.2 |
| NM_002842.4 | 20 | 3719 | NP_002833.4 | |
| XM_011527183.1 | 20 | 3740 | XP_011525485.1 | |
| XM_011527188.1 | 19 | 3689 | XP_011525490.1 | |
| XM_017027056.1 | 19 | 3727 | XP_016882545.1 | |
| XM_017027057.1 | 19 | 3715 | XP_016882546.1 | |
| XM_017027058.1 | 19 | 3760 | XP_016882547.1 | |
| XM_017027059.1 | 19 | 3679 | XP_016882548.1 | |
| XM_017027060.1 | 18 | 3679 | XP_016882549.1 | |
| XM_017027061.1 | 17 | 3193 | XP_016882550.1 | |
| XM_017027062.1 | 19 | 3635 | XP_016882551.1 | |
| XM_017027063.1 | 19 | 3628 | XP_016882552.1 | |
| AA573849.1 | 1 | 217 | ||
| D15049.2 | 20 | 3693 | BAA03645.2 |
More Information
Set Membership: |
|

Genomic Map