Assay Details
Target Gene Details
Entrez Gene ID: | 1584 |
Gene Name: | cytochrome P450 family 11 subfamily B member 1 |
Gene Aliases: |
CPN1, CYP11B, FHI, P450C11 |
Location: |
Chr.8:142872354-142879839 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CYP11B1 | NM_000497.3 | NP_000488.3 | ||
| NM_001026213.1 | NP_001021384.1 | |||
| XM_011516874.2 | XP_011515176.1 | |||
| XM_011516875.1 | 10 | 1439 | XP_011515177.1 | |
| XM_017013146.1 | 4 | 857 | XP_016868635.1 | |
| AF478474.1 | AAL84813.1 | |||
| AK094090.1 | BAC04283.1 | |||
| BC096285.3 | AAH96285.1 | |||
| BC096286.3 | AAH96286.1 | |||
| BC096287.3 | AAH96287.1 | |||
| BX647738.1 | ||||
| X55764.1 | CAA39290.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv482123 | Chr.8:142872359 - 142879820 on Build GRCh38 | Gain |
|
| esv2661137 | Chr.8:142256294 - 142935448 on Build GRCh38 | Deletion |
|
| nsv470246 | Chr.8:142872807 - 142926765 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map