Assay Details
Target Gene Details
Entrez Gene ID: | 153241 |
Gene Name: | centrosomal protein 120 |
Gene Aliases: |
CCDC100, SRTD13 |
Location: |
Chr.5:123344885-123423592 on Build GRCh38 |
Assay Gene Location: | Within Exon 21 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CEP120 | NM_001166226.1 | 20 | 3955 | NP_001159698.1 |
| NM_153223.3 | 21 | 4036 | NP_694955.2 | |
| XM_005271901.4 | 20 | 3817 | XP_005271958.1 | |
| XM_011543185.2 | 20 | 4370 | XP_011541487.1 | |
| XM_011543186.2 | 17 | 3649 | XP_011541488.1 | |
| XM_017009085.1 | 17 | 3514 | XP_016864574.1 | |
| BX648687.1 | 11 | 2429 | CAH10561.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1161297 | Chr.5:123340647 - 123372682 on Build GRCh38 | Duplication |
|
| nsv830472 | Chr.5:123240869 - 123416726 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map