Assay Details
Target Gene Details
Entrez Gene ID: | 100130935 |
Gene Name: | CSAG family member 4 (pseudogene) |
Gene Aliases: |
- |
Location: |
Chr.X:152733827-152740985 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CSAG4 | NR_073432.1 |
Target Gene Details
Entrez Gene ID: | 4111 |
Gene Name: | MAGE family member A12 |
Gene Aliases: |
CT1.12, MAGE12 |
Location: |
Chr.X:152733779-152737669 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MAGEA12 | NM_001166386.3 | 3 | 628 | NP_001159858.1 |
| NM_001166387.3 | 3 | 536 | NP_001159859.1 | |
| NM_005367.6 | 2 | 430 | NP_005358.2 | |
| AK314168.1 | 3 | 628 | ||
| BC003408.2 | 2 | 418 | AAH03408.1 | |
| BQ436888.1 | 2 | 430 | ||
| BT007108.1 | 1 | 274 | AAP35772.1 | |
| CF123053.1 | ||||
| CR541775.1 | 1 | 274 | CAG46574.1 |
More Information
Additional Information:
Set Membership: |
|

Genomic Map