Assay Details
Target Gene Details
Entrez Gene ID: | 7511 |
Gene Name: | X-prolyl aminopeptidase 1 |
Gene Aliases: |
APP1, SAMP, XPNPEP, XPNPEPL, XPNPEPL1 |
Location: |
Chr.10:109864766-109923553 on Build GRCh38 |
Assay Gene Location: | Within Exon 24 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| XPNPEP1 | NM_001167604.1 | 20 | 2349 | NP_001161076.1 |
| NM_001324128.1 | 19 | 2268 | NP_001311057.1 | |
| NM_001324131.1 | 21 | 2465 | NP_001311060.1 | |
| NM_001324132.1 | 20 | 2332 | NP_001311061.1 | |
| NM_001324133.1 | 22 | 2479 | NP_001311062.1 | |
| NM_001324134.1 | 19 | 2252 | NP_001311063.1 | |
| NM_001324135.1 | 20 | 2370 | NP_001311064.1 | |
| NM_001324136.1 | 21 | 2636 | NP_001311065.1 | |
| NM_020383.3 | 21 | 2421 | NP_065116.3 | |
| XM_011540134.1 | 20 | 2241 | XP_011538436.1 | |
| XM_017016613.1 | 20 | 2271 | XP_016872102.1 | |
| XM_017016614.1 | 22 | 2404 | XP_016872103.1 | |
| XM_017016615.1 | 20 | 2288 | XP_016872104.1 | |
| AF195530.1 | 20 | 2248 | AAF97866.1 | |
| AK093693.1 | 19 | 2226 | ||
| AK094900.1 | 1 | 2001 | ||
| AK095447.1 | 6 | 2467 | ||
| AL833411.1 | 20 | 2255 | CAD38640.1 | |
| BC005126.2 | 20 | 2210 | AAH05126.1 | |
| BC007579.1 | 21 | 2312 | AAH07579.1 | |
| BC013417.2 | 17 | 1922 | AAH13417.4 | |
| BX537682.1 | 17 | 4266 | CAH18706.2 |
More Information
Set Membership: |
|

Genomic Map