Assay Details
Target Gene Details
Entrez Gene ID: | 9131 |
Gene Name: | apoptosis inducing factor, mitochondria associated 1 |
Gene Aliases: |
AIF, CMT2D, CMTX4, COWCK, COXPD6, DFNX5, NADMR, NAMSD, PDCD8 |
Location: |
Chr.X:130129362-130165887 on Build GRCh38 |
Assay Gene Location: | Within Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AIFM1 | NM_001130846.3 | 7 | 1788 | NP_001124318.2 |
| NM_001130847.3 | 17 | 2334 | NP_001124319.1 | |
| NM_004208.3 | 16 | 2131 | NP_004199.1 | |
| NM_145812.2 | 16 | 2119 | NP_665811.1 | |
| NR_132647.1 | 16 | 2191 | ||
| AF131759.1 | 13 | 1483 | AAD20036.1 | |
| AI521711.1 | 1 | 155 | ||
| AK000775.1 | 16 | 1967 | ||
| AL049703.1 | 16 | 1984 | CAB41267.1 | |
| AL049704.1 | 8 | 1063 | CAB41268.1 | |
| BC111065.1 | 16 | 2062 | AAI11066.1 | |
| BC139738.1 | ||||
| DQ016496.2 | 7 | 1788 | AAY84737.1 | |
| DQ016498.1 | 17 | 2111 | ||
| DQ016500.1 | 16 | 1968 |
Target Gene Details
Entrez Gene ID: | 9363 |
Gene Name: | RAB33A, member RAS oncogene family |
Gene Aliases: |
RabS10 |
Location: |
Chr.X:130110633-130184872 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RAB33A | XM_017029963.1 | XP_016885452.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv528290 | Chr.X:129635682 - 130429852 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map