Assay Details
Target Gene Details
Entrez Gene ID: | 65095 |
Gene Name: | KRI1 homolog |
Gene Aliases: |
- |
Location: |
Chr.19:10553085-10566031 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KRI1 | NM_023008.3 | NP_075384.3 | ||
| XM_011528190.2 | XP_011526492.1 | |||
| AK023011.1 | BAB14357.1 | |||
| AK093879.1 | BAC04240.1 | |||
| AK300780.1 | ||||
| AK300850.1 | ||||
| AL833189.1 | ||||
| AU280109.1 | ||||
| BC002890.2 | AAH02890.3 | |||
| BC112249.1 | AAI12250.1 | |||
| BC112251.1 | AAI12252.1 | |||
| BC143770.1 | ||||
| DA438802.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833744 | Chr.19:10458964 - 10635033 on Build GRCh38 | Loss |
|
| nsv833745 | Chr.19:10539132 - 10727920 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map