Assay Details
Target Gene Details
Entrez Gene ID: | 2803 |
Gene Name: | golgin A4 |
Gene Aliases: |
CRPF46, GCP2, GOLG, MU-RMS-40.18, p230 |
Location: |
Chr.3:37243191-37366879 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GOLGA4 | NM_001172713.1 | 2 | 466 | NP_001166184.1 |
| NM_002078.4 | 2 | 466 | NP_002069.2 | |
| XM_005265069.3 | 2 | 386 | XP_005265126.1 | |
| XM_005265070.3 | 2 | 386 | XP_005265127.1 | |
| XM_005265071.3 | 2 | 386 | XP_005265128.1 | |
| XM_005265072.3 | 2 | 386 | XP_005265129.1 | |
| XM_005265073.1 | 2 | 386 | XP_005265130.1 | |
| XM_005265074.1 | 2 | 386 | XP_005265131.1 | |
| XM_005265075.1 | 2 | 388 | XP_005265132.1 | |
| XM_006713110.2 | 2 | 386 | XP_006713173.1 | |
| XM_017006184.1 | 2 | 386 | XP_016861673.1 | |
| XM_017006185.1 | 2 | 876 | XP_016861674.1 | |
| XM_017006186.1 | 2 | 55 | XP_016861675.1 | |
| XM_017006187.1 | 2 | 386 | XP_016861676.1 | |
| XM_017006188.1 | 2 | 388 | XP_016861677.1 | |
| AB209693.1 | 2 | 381 | BAD92930.1 | |
| AK290550.1 | 2 | 388 | ||
| AK300192.1 | 2 | 386 | ||
| AK307429.1 | 2 | 386 | ||
| BC045543.1 | AAH45543.1 | |||
| BC050444.1 | 2 | 410 | AAH50444.1 | |
| U41740.1 | 2 | 391 | AAC50434.1 | |
| X82834.1 | 2 | 313 | CAA58041.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834665 | Chr.3:37180973 - 37368088 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map