Assay Details
Target Gene Details
Entrez Gene ID: | 10825 |
Gene Name: | neuraminidase 3 |
Gene Aliases: |
SIAL3 |
Location: |
Chr.11:74988905-75007698 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NEU3 | NM_006656.5 | NP_006647.3 | ||
| AK290442.1 | ||||
| Y18563.1 | CAB96131.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832210 | Chr.11:74851737 - 75021088 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map