Assay Details
Target Gene Details
Entrez Gene ID: | 171484 |
Gene Name: | family with sequence similarity 9 member C |
Gene Aliases: |
TEX39C |
Location: |
Chr.X:13035617-13044798 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM9C | NM_174901.5 | NP_777561.1 | ||
| AF494345.1 | AAN07164.1 | |||
| AK313776.1 | ||||
| BX647580.1 | CAI46094.1 | |||
| BX648111.1 | CAI46034.1 | |||
| BX648919.1 | CAI46007.1 | |||
| DA958431.1 |
Target Gene Details
Entrez Gene ID: | 105373133 |
Gene Name: | uncharacterized LOC105373133 |
Gene Aliases: |
- |
Location: |
Chr.X:13044544-13086975 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC105373133 | XM_011545614.2 | XP_011543916.1 |
More Information
Set Membership: |
|

Genomic Map