Assay Details
Target Gene Details
Entrez Gene ID: | 9936 |
Gene Name: | CD302 molecule |
Gene Aliases: |
BIMLEC, CLEC13A, DCL-1, DCL1 |
Location: |
Chr.2:159768628-159798255 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CD302 | NM_001198763.1 | NP_001185692.1 | ||
| NM_001198764.1 | NP_001185693.1 | |||
| NM_014880.4 | NP_055695.2 | |||
| AF499000.1 | AAN77050.1 | |||
| AK291279.1 | ||||
| AK304422.1 | ||||
| AY314007.1 | AAP79900.1 | |||
| BC020646.1 | AAH20646.1 | |||
| D14664.1 | BAA03498.1 | |||
| DB217404.1 | ||||
| DB460994.1 |
Target Gene Details
Entrez Gene ID: | 100526664 |
Gene Name: | LY75-CD302 readthrough |
Gene Aliases: |
- |
Location: |
Chr.2:159768628-159904756 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 36 - Exon 36 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LY75-CD302 | NM_001198759.1 | NP_001185688.1 | ||
| NM_001198760.1 | NP_001185689.1 | |||
| AY184222.1 | AAN85434.1 | |||
| AY314006.1 | AAP79899.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834436 | Chr.2:159765830 - 159941264 on Build GRCh38 | Loss |
|
| nsv1010774 | Chr.2:159012247 - 160175703 on Build GRCh38 | Gain |
|
| nsv470497 | Chr.2:159694632 - 159836250 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map