Assay Details
Target Gene Details
Entrez Gene ID: | 90233 |
Gene Name: | zinc finger protein 551 |
Gene Aliases: |
- |
Location: |
Chr.19:57681969-57689811 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZNF551 | NM_001270938.1 | 3 | 2551 | NP_001257867.1 |
| NM_138347.4 | 3 | 2673 | NP_612356.2 | |
| NR_073102.1 | 2 | 2549 | ||
| AK091273.1 | BAC03625.1 | |||
| BC005868.2 | 3 | 2605 | AAH05868.1 | |
| BX538151.1 | 2 | 2519 | CAD98037.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv580402 | Chr.19:57622904 - 57704099 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map