Assay Details
Target Gene Details
Entrez Gene ID: | 4253 |
Gene Name: | CTAGE family member 5, ER export factor |
Gene Aliases: |
MEA6, MGEA, MGEA11, MGEA6 |
Location: |
Chr.14:39265264-39351193 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CTAGE5 | NM_203354.2 | NP_976229.1 | ||
| XM_011536778.2 | XP_011535080.1 | |||
| XM_011536785.2 | XP_011535087.1 | |||
| XM_017021316.1 | XP_016876805.1 | |||
| XM_017021322.1 | XP_016876811.1 | |||
| XM_017021324.1 | XP_016876813.1 | |||
| XM_017021327.1 | XP_016876816.1 | |||
| AK298935.1 | ||||
| BX640994.1 | CAE45997.1 | |||
| BX648014.1 | ||||
| CD512382.1 | ||||
| DA072098.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1053133 | Chr.14:39188331 - 39301525 on Build GRCh38 | Gain |
|
| esv3584293 | Chr.14:39191534 - 39339749 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map