Assay Details
Target Gene Details
Entrez Gene ID: | 81470 |
Gene Name: | olfactory receptor family 2 subfamily G member 2 |
Gene Aliases: |
OR1-32 |
Location: |
Chr.1:247588360-247589313 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| OR2G2 | NM_001001915.1 | 1 | 175 | NP_001001915.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2759014 | Chr.1:247551283 - 248159014 on Build GRCh38 | Gain+Loss |
|
| esv33129 | Chr.1:246911128 - 248165533 on Build GRCh38 | Gain+Loss |
|
| nsv1006136 | Chr.1:247539376 - 247637063 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map