Assay Details
Target Gene Details
Entrez Gene ID: | 80271 |
Gene Name: | inositol-trisphosphate 3-kinase C |
Gene Aliases: |
IP3-3KC, IP3KC |
Location: |
Chr.19:40717103-40740860 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ITPKC | NM_025194.2 | 3 | 1383 | NP_079470.1 |
| XM_017027324.1 | 4 | 567 | XP_016882813.1 | |
| AJ290975.1 | 3 | 1350 | CAC40815.1 | |
| AK024596.1 | 3 | 418 | ||
| BC060788.1 | 3 | 1383 | AAH60788.1 | |
| D38169.1 | 3 | 1114 | BAA22524.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1160619 | Chr.19:40706352 - 40763204 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map