Assay Details
Target Gene Details
Entrez Gene ID: | 57526 |
Gene Name: | protocadherin 19 |
Gene Aliases: |
EFMR, EIEE9 |
Location: |
Chr.X:100291644-100410273 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PCDH19 | NM_001105243.1 | 5 | 6409 | NP_001098713.1 |
| NM_001184880.1 | 6 | 6550 | NP_001171809.1 | |
| NM_020766.2 | 5 | 6406 | NP_065817.2 | |
| XM_011530997.2 | 6 | 5516 | XP_011529299.1 | |
| AB037734.1 | 5 | 2624 | BAA92551.1 | |
| BC136628.1 | 5 | 3290 | ||
| CR749278.1 | 5 | 3287 | CAH18133.1 | |
| EF676096.1 | 6 | 6550 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv7007 | Chr.X:100226294 - 100297292 on Build GRCh38 | Deletion |
|
| esv2752340 | Chr.X:99231803 - 100513048 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map