Assay Details
Target Gene Details
Entrez Gene ID: | 221914 |
Gene Name: | glypican 2 |
Gene Aliases: |
- |
Location: |
Chr.7:100169606-100177426 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GPC2 | NM_152742.2 | NP_689955.1 | ||
| AK096281.1 | BAC04745.1 | |||
| AL834418.1 | CAD39080.1 | |||
| BC027972.1 | AAH27972.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv5867 | Chr.7:100169151 - 100202132 on Build GRCh38 | Deletion |
|
| nsv607931 | Chr.7:100067194 - 100198523 on Build GRCh38 | Loss |
|
| esv2734888 | Chr.7:99975623 - 101290285 on Build GRCh38 | Deletion |
|
| nsv428180 | Chr.7:100153358 - 100363277 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map