Assay Details
Target Gene Details
Entrez Gene ID: | 59 |
Gene Name: | actin, alpha 2, smooth muscle, aorta |
Gene Aliases: |
AAT6, ACTSA, MYMY5 |
Location: |
Chr.10:88935074-88991397 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 9 - Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ACTA2 | NM_001141945.2 | NP_001135417.1 | ||
| NM_001320855.1 | NP_001307784.1 | |||
| NM_001613.2 | NP_001604.1 | |||
| AK127229.1 | 1 | 4015 | ||
| AK300664.1 | ||||
| AK313294.1 | ||||
| AY692464.1 | ||||
| BC017554.2 | AAH17554.1 | |||
| BC093052.1 | AAH93052.1 | |||
| BQ029841.1 | ||||
| CR536518.1 | CAG38756.1 | |||
| J05192.1 | AAA51577.1 | |||
| X13839.1 | CAA32064.1 |
Target Gene Details
Entrez Gene ID: | 100132116 |
Gene Name: | ACTA2 antisense RNA 1 |
Gene Aliases: |
ZXF1, uc001kfo.1 |
Location: |
Chr.10:88932684-88939974 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ACTA2-AS1 | NR_125373.1 | 3 | 993 | |
| AK093340.1 | 3 | 993 | ||
| AX748062.1 | 3 | 993 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv948130 | Chr.10:88935211 - 88935821 on Build GRCh38 | Duplication |
|
| nsv831943 | Chr.10:88905079 - 89106077 on Build GRCh38 | Gain |
|
| nsv831942 | Chr.10:88804940 - 88970595 on Build GRCh38 | Gain |
|
| esv3624144 | Chr.10:88876937 - 89011636 on Build GRCh38 | Gain |
|
| esv3624139 | Chr.10:88724980 - 89003307 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map