Assay Details
Target Gene Details
Entrez Gene ID: | 55300 |
Gene Name: | phosphatidylinositol 4-kinase type 2 beta |
Gene Aliases: |
PI4KIIB, PIK42B |
Location: |
Chr.4:25234031-25279209 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PI4K2B | NM_018323.3 | 1 | 118 | NP_060793.2 |
| XM_005248174.1 | 1 | 116 | XP_005248231.1 | |
| AF411320.1 | AAL04154.1 | |||
| AK001967.1 | 1 | 106 | BAA92006.1 | |
| AK128622.1 | 1 | 116 | ||
| AK296302.1 | ||||
| AY065990.1 | 1 | 96 | AAL47580.1 | |
| AY091514.1 | 1 | 105 | AAM12049.1 | |
| BC051749.1 | 1 | 118 | AAH51749.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3599959 | Chr.4:25219862 - 25235691 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map