Assay Details
Target Gene Details
Entrez Gene ID: | 57703 |
Gene Name: | CWC22 homolog, spliceosome-associated protein |
Gene Aliases: |
EIF4GL, NCM, fSAPb |
Location: |
Chr.2:179944876-180007359 on Build GRCh38 |
Assay Gene Location: | Within Exon 20 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CWC22 | NM_020943.2 | 19 | 2282 | NP_065994.1 |
| XM_005246726.2 | 19 | 2385 | XP_005246783.1 | |
| AB046824.1 | 18 | 2101 | ||
| AK025635.1 | 19 | 2138 | BAB15197.1 | |
| AK026978.1 | 12 | 1239 | BAB15612.1 | |
| AK123647.1 | 6 | 1239 | ||
| BC016651.1 | 18 | 2106 | ||
| BC031216.1 | 19 | 2282 | AAH31216.1 | |
| BC053573.1 | 16 | 1804 | AAH53573.1 | |
| BC057826.1 | 19 | 2140 | AAH57826.1 | |
| BC093952.1 | 18 | 2004 | AAH93952.1 | |
| BC093954.1 | 18 | 2004 | AAH93954.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv521134 | Chr.2:179881314 - 179983761 on Build GRCh38 | Gain |
|
| nsv1006598 | Chr.2:179902256 - 180058324 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map