Assay Details
Target Gene Details
Entrez Gene ID: | 343099 |
Gene Name: | coiled-coil domain containing 18 |
Gene Aliases: |
NY-SAR-41 |
Location: |
Chr.1:93179802-93279037 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CCDC18 | NM_001306076.1 | 2 | 236 | NP_001293005.1 |
| NM_206886.4 | 2 | 236 | NP_996769.3 | |
| XM_011541361.2 | 2 | 445 | XP_011539663.2 | |
| XM_011541372.2 | 2 | 445 | XP_011539674.2 | |
| XM_017001154.1 | 2 | 445 | XP_016856643.1 | |
| XM_017001155.1 | 2 | 445 | XP_016856644.1 | |
| XM_017001156.1 | 2 | 415 | XP_016856645.1 | |
| XM_017001157.1 | 2 | 445 | XP_016856646.1 | |
| XM_017001158.1 | 2 | 445 | XP_016856647.1 | |
| XM_017001159.1 | 2 | 445 | XP_016856648.1 | |
| XM_017001160.1 | 2 | 445 | XP_016856649.1 | |
| XM_017001161.1 | 2 | 445 | XP_016856650.1 | |
| XM_017001162.1 | 2 | 158 | XP_016856651.1 | |
| XM_017001163.1 | 2 | 712 | XP_016856652.1 | |
| XM_017001164.1 | 2 | 256 | XP_016856653.1 | |
| XM_017001165.1 | 2 | 445 | XP_016856654.1 | |
| XM_017001166.1 | 1 | 69 | XP_016856655.1 | |
| XM_017001167.1 | XP_016856656.1 | |||
| XM_017001169.1 | 2 | 445 | XP_016856658.1 | |
| BC056667.1 | 2 | 232 | AAH56667.1 | |
| BX099893.1 | 2 | 206 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830625 | Chr.1:93100821 - 93269689 on Build GRCh38 | Loss |
|
| nsv830614 | Chr.1:93000421 - 93213143 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map