Assay Details
Target Gene Details
Entrez Gene ID: | 5352 |
Gene Name: | procollagen-lysine,2-oxoglutarate 5-dioxygenase 2 |
Gene Aliases: |
BRKS2, LH2, TLH |
Location: |
Chr.3:146069436-146161495 on Build GRCh38 |
Assay Gene Location: | Within Exon 21 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PLOD2 | NM_000935.2 | 18 | 2503 | NP_000926.2 |
| NM_182943.2 | 19 | 2566 | NP_891988.1 | |
| XM_005247535.3 | 20 | 2123 | XP_005247592.1 | |
| XM_017006625.1 | 20 | 2077 | XP_016862114.1 | |
| AB209879.1 | 18 | 2175 | BAD93116.1 | |
| AK125700.1 | 9 | 1549 | ||
| AK295084.1 | 19 | 2157 | ||
| AL832238.1 | 8 | 725 | ||
| BC016834.1 | 5 | 416 | AAH16834.1 | |
| BC037169.1 | 19 | 2237 | AAH37169.1 | |
| U84573.1 | 18 | 1997 | AAB58363.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv525829 | Chr.3:145896530 - 146145992 on Build GRCh38 | Loss |
|
| nsv4046 | Chr.3:145975462 - 146162402 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map