Assay Details
Target Gene Details
Entrez Gene ID: | 284106 |
Gene Name: | CDGSH iron sulfur domain 3 |
Gene Aliases: |
Miner2 |
Location: |
Chr.17:38730257-38735605 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CISD3 | NM_001136498.1 | 4 | 1060 | NP_001129970.1 |
| AK097047.1 | 3 | 1311 | ||
| AK098433.1 | 2 | 841 |
Target Gene Details
Entrez Gene ID: | 7703 |
Gene Name: | polycomb group ring finger 2 |
Gene Aliases: |
MEL-18, RNF110, ZNF144 |
Location: |
Chr.17:38733897-38749817 on Build GRCh38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PCGF2 | NM_007144.2 | 11 | 2472 | NP_009075.1 |
| XM_005257640.2 | 12 | 2931 | XP_005257697.1 | |
| XM_005257641.4 | 10 | 3153 | XP_005257698.1 | |
| XM_005257642.3 | 10 | 3550 | XP_005257699.1 | |
| XM_017025016.1 | 12 | 3491 | XP_016880505.1 | |
| CA421781.1 | 1 | 151 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833438 | Chr.17:38593441 - 38821266 on Build GRCh38 | Loss |
|
| nsv470585 | Chr.17:38701569 - 38760346 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map