Assay Details
Target Gene Details
Entrez Gene ID: | 59345 |
Gene Name: | G protein subunit beta 4 |
Gene Aliases: |
CMTD1F |
Location: |
Chr.3:179396088-179451583 on Build GRCh38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GNB4 | NM_021629.3 | 10 | 1922 | NP_067642.1 |
| XM_005247692.2 | 10 | 1843 | XP_005247749.1 | |
| XM_006713721.2 | 10 | 1853 | XP_006713784.1 | |
| AK001890.1 | 10 | 1922 | ||
| AK022599.1 | 1 | 616 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv4139 | Chr.3:179384796 - 179418003 on Build GRCh38 | Insertion |
|
| nsv829806 | Chr.3:179234575 - 179436585 on Build GRCh38 | Gain |
|
| dgv4999n100 | Chr.3:179227018 - 179528391 on Build GRCh38 | Gain |
|
| nsv592650 | Chr.3:179250846 - 179534475 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs73045155] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map