Assay Details
Target Gene Details
Entrez Gene ID: | 163351 |
Gene Name: | guanylate binding protein family member 6 |
Gene Aliases: |
- |
Location: |
Chr.1:89363877-89388160 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GBP6 | NM_001320257.1 | NP_001307186.1 | ||
| NM_198460.2 | 2 | 347 | NP_940862.2 | |
| XM_011540835.2 | 2 | 176 | XP_011539137.1 | |
| AK131356.1 | 2 | 331 | BAD18509.1 | |
| AK290303.1 | 2 | 167 | ||
| AK299408.1 | 2 | 162 | ||
| AK299466.1 | ||||
| AK303987.1 | 2 | 178 | ||
| AK315834.1 | 2 | 165 | ||
| BC131713.1 | 2 | 125 | ||
| BX537949.1 | 2 | 166 | CAD97917.1 | |
| BX647907.1 | 2 | 164 | ||
| DA437660.1 | 2 | 345 | ||
| DA447005.1 | 2 | 331 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3586760 | Chr.1:89283608 - 89381169 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map