Assay Details
Target Gene Details
Entrez Gene ID: | 9908 |
Gene Name: | G3BP stress granule assembly factor 2 |
Gene Aliases: |
- |
Location: |
Chr.4:75642769-75724525 on Build GRCh38 |
Assay Gene Location: | Within Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| G3BP2 | NM_012297.4 | 12 | 2186 | NP_036429.2 |
| NM_203504.2 | 11 | 2222 | NP_987100.1 | |
| NM_203505.2 | 12 | 2321 | NP_987101.1 | |
| XM_005263382.2 | 12 | 2295 | XP_005263439.1 | |
| XM_005263383.3 | 13 | 2260 | XP_005263440.1 | |
| XM_011532441.2 | 12 | 2181 | XP_011530743.1 | |
| XM_017008876.1 | 12 | 2169 | XP_016864365.1 | |
| XM_017008877.1 | 12 | 2348 | XP_016864366.1 | |
| XM_017008878.1 | 11 | 2070 | XP_016864367.1 | |
| XM_017008879.1 | 11 | 2096 | XP_016864368.1 | |
| AB014560.1 | 12 | 2141 | ||
| AK291772.1 | ||||
| AK291786.1 | ||||
| BC011731.2 | 11 | 2140 | AAH11731.1 |
More Information
Set Membership: |
|

Genomic Map