Assay Details
Target Gene Details
Entrez Gene ID: | 941 |
Gene Name: | CD80 molecule |
Gene Aliases: |
B7, B7-1, B7.1, BB1, CD28LG, CD28LG1, LAB7 |
Location: |
Chr.3:119523909-119559709 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CD80 | NM_005191.3 | NP_005182.1 | ||
| XM_011513327.2 | XP_011511629.1 | |||
| AK292129.1 | ||||
| AY197777.1 | AAO39208.1 | |||
| AY197778.1 | AAO39209.1 | |||
| BC042665.1 | AAH42665.1 | |||
| M27533.1 | 6 | 1209 | AAA36045.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv829695 | Chr.3:119460043 - 119649149 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map