Assay Details
Target Gene Details
Entrez Gene ID: | 3157 |
Gene Name: | 3-hydroxy-3-methylglutaryl-CoA synthase 1 |
Gene Aliases: |
HMGCS |
Location: |
Chr.5:43287470-43313512 on Build GRCh38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HMGCS1 | NM_001098272.2 | 11 | 1923 | NP_001091742.1 |
| NM_001324219.1 | 10 | 1910 | NP_001311148.1 | |
| NM_001324220.1 | 11 | 1943 | NP_001311149.1 | |
| NM_001324222.1 | 10 | 1797 | NP_001311151.1 | |
| NM_001324223.1 | 9 | 1738 | NP_001311152.1 | |
| NM_001324224.1 | 10 | 1817 | NP_001311153.1 | |
| NM_002130.7 | 10 | 1864 | NP_002121.4 | |
| XM_011514036.2 | 8 | 1729 | XP_011512338.1 | |
| XM_017009412.1 | 11 | 2003 | XP_016864901.1 | |
| AK095492.1 | 11 | 1731 | BAC04559.1 | |
| BC000297.2 | 11 | 1802 | AAH00297.2 | |
| BC083514.1 | 10 | 1762 | AAH83514.1 | |
| L25798.1 | AAA62411.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv508352 | Chr.5:43237728 - 43390960 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map