Assay Details
Target Gene Details
Entrez Gene ID: | 55631 |
Gene Name: | leucine rich repeat containing 40 |
Gene Aliases: |
dJ677H15.1 |
Location: |
Chr.1:70144802-70205678 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LRRC40 | NM_017768.4 | 7 | 1073 | NP_060238.3 |
| XM_005271013.1 | 6 | 900 | XP_005271070.1 | |
| XM_011541763.1 | 5 | 815 | XP_011540065.1 | |
| XM_017001695.1 | 6 | 821 | XP_016857184.1 | |
| XM_017001696.1 | 4 | 595 | XP_016857185.1 | |
| AK000338.1 | 5 | 583 | BAA91094.1 | |
| AK022951.1 | 7 | 987 | BAB14326.1 | |
| AK225337.1 | 5 | 583 | ||
| BC003407.1 | 7 | 873 | AAH03407.2 | |
| BC008586.1 | 7 | 971 | AAH08586.1 |
More Information
Set Membership: |
|

Genomic Map