Assay Details
Target Gene Details
Entrez Gene ID: | 8905 |
Gene Name: | adaptor related protein complex 1 sigma 2 subunit |
Gene Aliases: |
DC22, MRX59, MRXS21, MRXS5, MRXSF, PGS, SIGMA1B |
Location: |
Chr.X:15825806-15855014 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AP1S2 | NM_001272071.1 | 6 | 2031 | NP_001259000.1 |
| NM_003916.4 | 5 | 2022 | NP_003907.3 | |
| AF251295.1 | 5 | 1778 | AAG44595.1 | |
| AK125525.1 | ||||
| AK130075.1 | ||||
| BX537780.1 | CAD97839.1 |
More Information
Set Membership: |
|

Genomic Map