Assay Details
Target Gene Details
Entrez Gene ID: | 221443 |
Gene Name: | O-acyl-ADP-ribose deacylase 1 |
Gene Aliases: |
C6orf130, TARG1, dJ34B21.3 |
Location: |
Chr.6:41066792-41072449 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| OARD1 | NM_145063.2 | 6 | 896 | NP_659500.1 |
| AJ420538.1 | 6 | 1032 | ||
| AK290939.1 | 6 | 899 | ||
| BC011709.2 | 6 | 896 | AAH11709.1 | |
| BC021579.1 | 5 | 846 |
More Information
Set Membership: |
|

Genomic Map