Assay Details
Target Gene Details
Entrez Gene ID: | 91746 |
Gene Name: | YTH domain containing 1 |
Gene Aliases: |
YT521, YT521-B |
Location: |
Chr.4:68310387-68350106 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| YTHDC1 | NM_001031732.2 | 9 | 1642 | NP_001026902.1 |
| NM_133370.2 | 8 | 1588 | NP_588611.2 | |
| XM_005265706.2 | 9 | 1642 | XP_005265763.1 | |
| XM_005265707.2 | 9 | 1776 | XP_005265764.1 | |
| XM_005265708.2 | 8 | 1588 | XP_005265765.1 | |
| XM_017008832.1 | 9 | 1776 | XP_016864321.1 | |
| XM_017008833.1 | 8 | 1722 | XP_016864322.1 | |
| AB075846.1 | 6 | 548 | BAB85552.1 | |
| AK056430.1 | 8 | 1573 | BAB71181.1 | |
| AK098515.1 | 6 | 928 | ||
| AL832559.1 | 6 | 1107 | ||
| BC041119.1 | 9 | 1572 | AAH41119.1 | |
| BC053863.1 | 8 | 1253 | AAH53863.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1122086 | Chr.4:68303982 - 69012282 on Build GRCh38 | Deletion |
|
| nsv1073769 | Chr.4:68304281 - 68524582 on Build GRCh38 | Deletion |
|
| esv2759258 | Chr.4:68304361 - 69548454 on Build GRCh38 | Gain+Loss |
|
| nsv1119002 | Chr.4:68304882 - 68474682 on Build GRCh38 | Deletion |
|
| nsv594506 | Chr.4:68183925 - 68357339 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map