Assay Details
Target Gene Details
Entrez Gene ID: | 5978 |
Gene Name: | RE1 silencing transcription factor |
Gene Aliases: |
NRSF, WT6, XBR |
Location: |
Chr.4:56907876-56935847 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 4 - Intron 5 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv5214n100 | Chr.4:56786612 - 56999637 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map