Assay Details
Target Gene Details
Entrez Gene ID: | 8861 |
Gene Name: | LIM domain binding 1 |
Gene Aliases: |
CLIM-2, CLIM2, LDB-1, NLI |
Location: |
Chr.10:102102100-102120473 on Build GRCh38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LDB1 | NM_001113407.2 | 11 | 1838 | NP_001106878.1 |
| NM_001321612.1 | 11 | 1832 | NP_001308541.1 | |
| NM_003893.4 | 11 | 2011 | NP_003884.1 | |
| XM_017016867.1 | XP_016872356.1 | |||
| XM_017016868.1 | XP_016872357.1 | |||
| XM_017016869.1 | 7 | 1129 | XP_016872358.1 | |
| AF064491.1 | 11 | 1680 | AAC28341.1 | |
| AF068652.1 | 11 | 1651 | AAC77818.1 | |
| BC000482.2 | 11 | 1990 | AAH00482.1 | |
| BC009246.2 | 11 | 1990 | AAH09246.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs114758555] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map