Assay Details
Target Gene Details
Entrez Gene ID: | 130940 |
Gene Name: | coiled-coil domain containing 148 |
Gene Aliases: |
- |
Location: |
Chr.2:158171356-158456753 on Build GRCh38 |
Assay Gene Location: | Within Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CCDC148 | NM_001301684.1 | 12 | 1734 | NP_001288613.1 |
| NM_138803.3 | 14 | 1990 | NP_620158.3 | |
| XM_005246319.3 | 15 | 1877 | XP_005246376.1 | |
| XM_011510638.2 | 16 | 1917 | XP_011508940.1 | |
| XM_017003392.1 | 15 | 2029 | XP_016858881.1 | |
| XM_017003393.1 | 15 | 1712 | XP_016858882.1 | |
| XM_017003394.1 | 14 | 1862 | XP_016858883.1 | |
| XM_017003395.1 | 14 | 1925 | XP_016858884.1 | |
| XM_017003396.1 | 13 | 1886 | XP_016858885.1 | |
| XM_017003397.1 | 11 | 1363 | XP_016858886.1 | |
| XM_017003398.1 | 9 | 1358 | XP_016858887.1 | |
| AF458588.1 | 14 | 1939 | AAM49716.1 | |
| AK123220.1 | 9 | 1219 | ||
| BC105928.1 | 13 | 1703 | AAI05929.1 | |
| BC107596.1 | 12 | 1547 | AAI07597.1 | |
| BC107597.1 | 14 | 1802 | AAI07598.1 |
Target Gene Details
Entrez Gene ID: | 554201 |
Gene Name: | CCDC148 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.2:158166650-158236169 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CCDC148-AS1 | NR_038850.1 |
More Information
Set Membership: |
|

Genomic Map